Learn More
TARGETMOL CHEMICALS INC Biotin LC hydrazide 50MG
SDP
Catalog No. 502878359
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Biotin LC hydrazide (BACH) is a modification reagent of long-chain protein. Biotin LC hydrazide transforms periodate-oxidized glycoproteins and can be used to target glycoproteins and glycans. Purity 99.4%
Catalog No. 50-287-8359 Supplier TARGETMOL CHEMICALS INC Supplier No. T1921050MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Biotin LC hydrazide (BACH) is a modification reagent of long-chain protein. Biotin LC hydrazide transforms periodate-oxidized glycoproteins and can be used to target glycoproteins and glycans. Purity 99.4%