Learn More
Medchemexpress LLC Biotin LC hydrazide (biotinamidocaproyl hydrazide) | 109276-34-8 | MFCD00077659 | 99.8% | 371.50 g/mol | C16H29N5O3S | 100 MG
SDP
Catalog No. 5000247944
Encompass_Preferred
Biotin LC hydrazide is a long-chain biotinylation reagent that reacts with periodate-oxidized glycoproteins and glycans to introduce biotin labels for detection, affinity capture, and conjugation workflows. It is carbohydrate-reactive and reported to be more sensitive than simple biotin hydrazide, improving labeling efficiency in biochemical assays.
  • Long-chain hydrazide reagent for labeling oxidized carbohydrates.
  • Reacts with periodate-oxidized glycoproteins and glycans.
  • Higher sensitivity than biotin hydrazide for improved detection.
  • Soluble in DMSO for solution preparation.
  • Suitable for biotinylation of proteins, glycans, and nucleic acids.
Catalog No. 50-002-47944 Supplier Medchemexpress LLC Supplier No. HY101885100MG
Biotin LC hydrazide is a long-chain biotinylation reagent that reacts with periodate-oxidized glycoproteins and glycans to introduce biotin labels for detection, affinity capture, and conjugation workflows. It is carbohydrate-reactive and reported to be more sensitive than simple biotin hydrazide, improving labeling efficiency in biochemical assays.
  • Long-chain hydrazide reagent for labeling oxidized carbohydrates.
  • Reacts with periodate-oxidized glycoproteins and glycans.
  • Higher sensitivity than biotin hydrazide for improved detection.
  • Soluble in DMSO for solution preparation.
  • Suitable for biotinylation of proteins, glycans, and nucleic acids.