Learn More
Cayman Chemical BIotIn-LC-LC-NHS 25mg
SDP
Catalog No. 502982161
Encompass_Preferred
A biotinylation reagent; contains an NHS moiety to facilitate coupling reactions and 30.5 Å spacer to decrease steric hindrance; has been used to biotinylate small molecules such as paclitaxel and coenzyme Q10
Catalog No. 50-298-2161 Supplier Cayman Chemical Supplier No. 1888725 MG
A biotinylation reagent; contains an NHS moiety to facilitate coupling reactions and 30.5 Å spacer to decrease steric hindrance; has been used to biotinylate small molecules such as paclitaxel and coenzyme Q10