Learn More
Cayman Chemical BIotIn-LC-LC-NHS 5mg
SDP
Catalog No. 502983486
Encompass_Preferred
A biotinylation reagent; contains an NHS moiety to facilitate coupling reactions and 30.5 Å spacer to decrease steric hindrance; has been used to biotinylate small molecules such as paclitaxel and coenzyme Q10
Catalog No. 50-298-3486 Supplier Cayman Chemical Supplier No. 188875 MG
A biotinylation reagent; contains an NHS moiety to facilitate coupling reactions and 30.5 Å spacer to decrease steric hindrance; has been used to biotinylate small molecules such as paclitaxel and coenzyme Q10