Learn More
Medchemexpress LLC Biotin-LC-LC-NHS | 89889-52-1 | 98.0% | 567.7 g/mol | C26H41N5O7S | 50 MG
SDP
Catalog No. 5000209329
Encompass_Preferred
Biotin-LC-LC-NHS is a long-chain N-hydroxysuccinimide (NHS) activated biotinylation reagent used to label proteins, antibodies, and small molecules. The LC-LC spacer reduces steric hindrance during conjugation, and the NHS ester reacts with primary amines to form stable amide bonds.
  • Provides long-chain spacer (LC-LC) to reduce steric hindrance.
  • Reactive NHS ester for amine-selective conjugation.
  • Suitable for labeling antibodies and small molecules.
  • Solid, white to light yellow appearance; store under inert gas and protect from light.
  • Soluble in DMSO; follow recommended solvent and storage guidance.
Catalog No. 50-002-09329 Supplier Medchemexpress LLC Supplier No. HYW04025450MG
Biotin-LC-LC-NHS is a long-chain N-hydroxysuccinimide (NHS) activated biotinylation reagent used to label proteins, antibodies, and small molecules. The LC-LC spacer reduces steric hindrance during conjugation, and the NHS ester reacts with primary amines to form stable amide bonds.
  • Provides long-chain spacer (LC-LC) to reduce steric hindrance.
  • Reactive NHS ester for amine-selective conjugation.
  • Suitable for labeling antibodies and small molecules.
  • Solid, white to light yellow appearance; store under inert gas and protect from light.
  • Soluble in DMSO; follow recommended solvent and storage guidance.