Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-lc-lc-nhs | 89889-52-1 | 98.0% | 567.70 | C26H41N5O7S | 25 MG
SDP

Catalog No. 5000208863
Encompass_Preferred

Biotin-LC-LC-NHS is an N-hydroxysuccinimide (NHS) ester biotinylation reagent with dual long-chain spacers. It covalently attaches biotin to primary amines on proteins, antibodies, and small molecules for labeling, detection, or affinity capture. Handle using appropriate safety precautions and store protected from light at -20°C.

  • NHS ester reactive toward primary amines.
  • Dual long-chain spacers reduce steric hindrance during labeling.
  • Molecular weight 567.70 and formula C26H41N5O7S for precise calculations.
  • Purity 98.0% for research-grade applications.
  • High solubility in DMSO (100 mg/mL) with recommended freshly opened DMSO.
  • Store at -20°C, protect from light, and handle under standard laboratory safety practices.

Catalog No. 50-002-08863 Supplier Medchemexpress LLC Supplier No. 25MGHYW040254
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-LC-LC-NHS is an N-hydroxysuccinimide (NHS) ester biotinylation reagent with dual long-chain spacers. It covalently attaches biotin to primary amines on proteins, antibodies, and small molecules for labeling, detection, or affinity capture. Handle using appropriate safety precautions and store protected from light at -20°C.

  • NHS ester reactive toward primary amines.
  • Dual long-chain spacers reduce steric hindrance during labeling.
  • Molecular weight 567.70 and formula C26H41N5O7S for precise calculations.
  • Purity 98.0% for research-grade applications.
  • High solubility in DMSO (100 mg/mL) with recommended freshly opened DMSO.
  • Store at -20°C, protect from light, and handle under standard laboratory safety practices.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.