Learn More
Medchemexpress LLC Biotin-LC-LC-NHS | 89889-52-1 | 98.0% | 567.70 g/mol | C26H41N5O7S | 100 MG
SDP
Catalog No. 5000209563
Encompass_Preferred
Biotin-LC-LC-NHS is an amino-reactive N-hydroxysuccinimide (NHS) ester used to biotinylate primary amines on proteins, antibodies, and small molecules. It contains an extended LC-LC spacer arm to improve accessibility of the biotin tag and is supplied as a solid for laboratory conjugation and cross-linking applications.
  • Reacts with primary amines for stable biotinylation.
  • Extended LC-LC spacer improves tag accessibility.
  • Provided as a solid, white to light yellow powder.
  • Soluble in DMSO at 100 mg/mL; compatible with common in vivo formulation vehicles.
  • Store protected from light under nitrogen at -20°C; in solution, store at -80°C for long-term storage.
Catalog No. 50-002-09563 Supplier Medchemexpress LLC Supplier No. HYW040254100MG
Biotin-LC-LC-NHS is an amino-reactive N-hydroxysuccinimide (NHS) ester used to biotinylate primary amines on proteins, antibodies, and small molecules. It contains an extended LC-LC spacer arm to improve accessibility of the biotin tag and is supplied as a solid for laboratory conjugation and cross-linking applications.
  • Reacts with primary amines for stable biotinylation.
  • Extended LC-LC spacer improves tag accessibility.
  • Provided as a solid, white to light yellow powder.
  • Soluble in DMSO at 100 mg/mL; compatible with common in vivo formulation vehicles.
  • Store protected from light under nitrogen at -20°C; in solution, store at -80°C for long-term storage.