Learn More
Vector Laboratories Biotin (Long Arm) Hydrazide, 50 mg
SDP
Catalog No. 502045706
Encompass
Specialty Products. Biotin (Long Arm) Hydrazide reacts with aldehyde groups and thus can be used to label carbohydrate groups present in a wide variety of macromolecules. The spacer arm in this product allows optimal accessibility of avidin or streptavidin conjugates to biotin. Incorporation of Biotin (Long Arm) Hydrazide involves a two-step process. First, the saccharide is treated with periodate (not included) that produces aldehyde groups on sugars containing unsubstituted vicinal hydroxyl groups (these are usually present at the non-reducing termini of the oligosaccharide). Next, Biotin (Long Arm) Hydrazide reacts with the aldehyde groups to produce a stable linkage between the saccharide and biotin. This reagent can also be used to couple biotin to carboxyl groups on proteins and other molecules using a carbodiimide coupling method, and to specifically label RNA with biotin at the 3' end.
Catalog No. 50-204-5706 Supplier Vector Laboratories Supplier No. SP110050
Specialty Products. Biotin (Long Arm) Hydrazide reacts with aldehyde groups and thus can be used to label carbohydrate groups present in a wide variety of macromolecules. The spacer arm in this product allows optimal accessibility of avidin or streptavidin conjugates to biotin. Incorporation of Biotin (Long Arm) Hydrazide involves a two-step process. First, the saccharide is treated with periodate (not included) that produces aldehyde groups on sugars containing unsubstituted vicinal hydroxyl groups (these are usually present at the non-reducing termini of the oligosaccharide). Next, Biotin (Long Arm) Hydrazide reacts with the aldehyde groups to produce a stable linkage between the saccharide and biotin. This reagent can also be used to couple biotin to carboxyl groups on proteins and other molecules using a carbodiimide coupling method, and to specifically label RNA with biotin at the 3' end.