Learn More
Raybiotech Inc Biotin Magnetic Beads
Catalog No. NC1586104
Encompass_Preferred
Superparamagnetic nanoparticles are coupled with Biotin and utilized in the magnetic separation and isolation of avidin and streptavidin-labeled components.
Catalog No. NC1586104 Supplier Raybiotech Inc Supplier No. 8011071
Superparamagnetic nanoparticles are coupled with Biotin and utilized in the magnetic separation and isolation of avidin and streptavidin-labeled components.