Learn More
Medchemexpress LLC Biotin-maleimide | 116919-18-7 | MFCD28397114 | 99.7% | 451.54 | C20H29N5O5S | 10mg
SDP
Catalog No. 5000268355
Encompass_Preferred
Biotin-maleimide is a biotinylated biochemical assay reagent which can be used as a thiol-specific probe Biotin-maleimide can also be used to prepare the biotin-conjugated polychelating agent[1]
Catalog No. 50-002-68355 Supplier Medchemexpress LLC Supplier No. HYW80875710MG
Biotin-maleimide is a biotinylated biochemical assay reagent which can be used as a thiol-specific probe Biotin-maleimide can also be used to prepare the biotin-conjugated polychelating agent[1]