Learn More
Bon Opus Biosciences Biotinylated Recombinant Mouse MerTK (C-hFc) 25ug
SDP
Supplier:  Bon Opus Biosciences BP110825UG
Encompass_Preferred
Biotinylated Recombinant Mouse MerTK (C-hFc) is produced by Human Cells. The target gene encoding Glu23 - Phe498 is expressed with a C-hFc tag. Accession# Q12866-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1108
Catalog No. 50-217-2704