Learn More
Biolegend Biotin (azide-free) anti-mouse/rat β-Amyloid, 1-16
Supplier:  Biolegend 805703
Encompass_Preferred
Biotin (azide-free) anti-mouse/rat β-Amyloid, 1-16 [M3.2]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: WB, ICC, ELISA; Size: 200 uL
Catalog No. 50-102-9601