Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (+)-Biotin N-hydroxysuccinimide ester, 98%

Catalog No. AC439300010
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439300010 1 g Glass bottle
AC439302500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439300010 Supplier Thermo Scientific Chemicals Supplier No. 439300010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 35013-72-0 | C14H19N3O5S | 341.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35013-72-0
Molecular Formula C14H19N3O5S
Molecular Weight (g/mol) 341.39
InChI Key YMXHPSHLTSZXKH-RVBZMBCESA-N
Synonym +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
PubChem CID 6710714
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 202.3°C to 202.7°C
Color White
Elemental Analysis 48.03 to 50.49% C, 12.00 to 12.62% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 341.39
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (+)-Biotin N-hydroxysuccinimide ester
Show More Show Less
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.