Learn More
Medchemexpress LLC Biotin-naphthylamine | 2375201-60-6 | MFCD34186905 | 98.3% | 426.57 | C23H30N4O2S | 5 MG
SDP
Catalog No. 5000283953
Encompass_Preferred
Biotin-naphthylamine is a biotinylated biochemical assay reagent used as a substrate for ascorbate peroxidase 2 (APEX2) and as a probe for labeling DNA and RNA. It is supplied as a solid, white to off-white material with high purity suitable for research applications.
  • Serves as a substrate for ascorbate peroxidase 2 (APEX2).
  • Useful probe for labeling DNA and RNA.
  • Supplied as a solid, white to off-white compound.
  • High purity (98.3%), suitable for biochemical assays.
  • Store protected from light and under nitrogen at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-83953 Supplier Medchemexpress LLC Supplier No. HYW6147535MG
Biotin-naphthylamine is a biotinylated biochemical assay reagent used as a substrate for ascorbate peroxidase 2 (APEX2) and as a probe for labeling DNA and RNA. It is supplied as a solid, white to off-white material with high purity suitable for research applications.
  • Serves as a substrate for ascorbate peroxidase 2 (APEX2).
  • Useful probe for labeling DNA and RNA.
  • Supplied as a solid, white to off-white compound.
  • High purity (98.3%), suitable for biochemical assays.
  • Store protected from light and under nitrogen at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).