Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-naphthylamine | 2375201-60-6 | MFCD34186905 | 98.3% | 426.57 g·mol⁻¹ | C23H30N4O2S | 10 MG
SDP

Catalog No. 5000284079
Encompass_Preferred

Biotin-naphthylamine is a biotinylated biochemical assay reagent that serves as a substrate for ascorbate peroxidase 2 (APEX2). It is used as a probe for labeling DNA and RNA in biochemical assays and is supplied as a high-purity solid intended for research use.

  • High purity (98.3%)
  • Solid form, convenient for storage and handling
  • Substrate for APEX2, enabling enzymatic labeling of nucleic acids
  • Molecular weight 426.57 g·mol⁻¹; CAS 2375201-60-6
  • Recommended storage: -20°C, protect from light, store under nitrogen

Catalog No. 50-002-84079 Supplier Medchemexpress LLC Supplier No. HYW61475310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-naphthylamine is a biotinylated biochemical assay reagent that serves as a substrate for ascorbate peroxidase 2 (APEX2). It is used as a probe for labeling DNA and RNA in biochemical assays and is supplied as a high-purity solid intended for research use.

  • High purity (98.3%)
  • Solid form, convenient for storage and handling
  • Substrate for APEX2, enabling enzymatic labeling of nucleic acids
  • Molecular weight 426.57 g·mol⁻¹; CAS 2375201-60-6
  • Recommended storage: -20°C, protect from light, store under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.