Learn More
Medchemexpress LLC Biotin-naphthylamine | 2375201-60-6 | MFCD34186905 | 98.3% | 426.57 g/mol | C23H30N4O2S | 25 MG
SDP
Catalog No. 5000284754
Encompass_Preferred
Biotin-naphthylamine is a biotinylated biochemical assay reagent used as a substrate for ascorbate peroxidase 2 (APEX2). It serves as a labeling probe for DNA and RNA in biochemical and molecular biology applications.
  • Biotinylated substrate for APEX2, enabling enzymatic labeling.
  • Suitable for DNA and RNA labeling applications.
  • High purity (98.3%) for reliable assay performance.
  • Molecular weight 426.57 g/mol; formula C23H30N4O2S.
  • Store protected from light and under nitrogen; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-84754 Supplier Medchemexpress LLC Supplier No. HYW61475325MG
Biotin-naphthylamine is a biotinylated biochemical assay reagent used as a substrate for ascorbate peroxidase 2 (APEX2). It serves as a labeling probe for DNA and RNA in biochemical and molecular biology applications.
  • Biotinylated substrate for APEX2, enabling enzymatic labeling.
  • Suitable for DNA and RNA labeling applications.
  • High purity (98.3%) for reliable assay performance.
  • Molecular weight 426.57 g/mol; formula C23H30N4O2S.
  • Store protected from light and under nitrogen; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).