Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin Nick Translation Mi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11745824910
Encompass
Convenient enzyme and nucleotide mixture.Nick translation utilizes a combination of DNase and DNA Polymerase to nick one strand of the DNA helix then incorporates labeled nucleotides as the polymerase examines or proofreads the nicked site. The molar ratio of Biotin-16-dUTP to dTTP is adjusted to ensure that every 20th to 25th nucleotide in the newly synthesized DNA is modified with biotin. This density of haptens in the DNA yields the highest sensitivity in the immunological detection reaction.Large plasmids cosmids and PCR fragments are all regularly used with this method and the DNA can be linearized or supercoiled. Individual templates produce consistent results in the standard 90-minutes reaction and result in an average probe length of 200 base pairs up to 500 base pairs.
Catalog No. 50-290-5203