Learn More
Aat Bioquest Biotin NTA (5 mg)
SDP
Supplier:  Aat Bioquest 3008
Encompass_Preferred
Biotin NTA BNTA a biotin-labeled nitrilotriacetic acid is widely used to detect histidine-tagged proteins immobilized on nitrocellulose membranes with the detection limite of 0.1 pmol. This catalog is for 5 mg.
Catalog No. 50-224-9380