Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-oxytocin (biotin-a-hypophamine; biotin-oxytocic hormone) | 99.3% | 1233.48 | C53H80N14O14S3 | 10 MG
SDP

Catalog No. 5000298753
Encompass_Preferred

Biotin-oxytocin is an N-terminally biotinylated form of the oxytocin peptide supplied for laboratory research use. The peptide retains the native disulfide-stabilized backbone while providing an N-terminal biotin tag for affinity capture, labeling, and detection in biochemical and cell-based assays. Supplied with supporting documentation including a datasheet and COA.

  • 99.3% purity.
  • Molecular weight 1233.48 Da.
  • Sequence: biotin-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 (disulfide Cys1-Cys6).
  • Available sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Supplied with datasheet, COA, and SDS.
  • Intended for research use only.

Catalog No. 50-002-98753 Supplier Medchemexpress LLC Supplier No. HY17571F10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-oxytocin is an N-terminally biotinylated form of the oxytocin peptide supplied for laboratory research use. The peptide retains the native disulfide-stabilized backbone while providing an N-terminal biotin tag for affinity capture, labeling, and detection in biochemical and cell-based assays. Supplied with supporting documentation including a datasheet and COA.

  • 99.3% purity.
  • Molecular weight 1233.48 Da.
  • Sequence: biotin-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 (disulfide Cys1-Cys6).
  • Available sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Supplied with datasheet, COA, and SDS.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.