Learn More
Medchemexpress LLC Biotin-PEG-amine (MW 2000) | 90.0% | 100 MG
Supplier: Medchemexpress LLC HY140654100MG
Biotin-PEG-amine (MW 2000) is a PEG-based PROTAC linker that facilitates the synthesis of PROTACs. PROTACs feature two ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
- PEG-based PROTAC linker
- Enables synthesis of PROTACs
- Utilizes the ubiquitin-proteasome system
- Selectively degrades target proteins