Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG-amine (MW 2000) | 90.0% | 250 MG
SDP

Catalog No. 5000352757
Encompass_Preferred

Biotin-PEG-amine (MW 2000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound is a white to off-white solid with an average molecular weight of 2000.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity of 90.0%
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-003-52757 Supplier Medchemexpress LLC Supplier No. 250MGHY140654
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG-amine (MW 2000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound is a white to off-white solid with an average molecular weight of 2000.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity of 90.0%
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.