Learn More
Medchemexpress LLC Biotin-PEG-azide | 956494-20-5 | 98.7% | 2000.00 | 100 MG
SDP
Catalog No. 5000281250
Encompass_Preferred
Biotin-PEG-azide (MW 2000) is a biotinylated polyethylene glycol derivative with a terminal azide functional group. Supplied as a powder with an average molecular weight of 2000, it is designed for use in bioconjugation workflows such as copper-catalyzed azide-alkyne cycloaddition and for affinity labeling or capture of biomolecules. Manufacturer datasheets provide purity, storage, and handling details.
Catalog No. 50-002-81250 Supplier Medchemexpress LLC Supplier No. HYW250928C100MG
Biotin-PEG-azide (MW 2000) is a biotinylated polyethylene glycol derivative with a terminal azide functional group. Supplied as a powder with an average molecular weight of 2000, it is designed for use in bioconjugation workflows such as copper-catalyzed azide-alkyne cycloaddition and for affinity labeling or capture of biomolecules. Manufacturer datasheets provide purity, storage, and handling details.