Learn More
Medchemexpress LLC Biotin-PEG-azide | 956494-20-5 | MFCD21363313 | 95.0% | 100 MG
SDP
Catalog No. 5000281632
Encompass_Preferred
Biotin-PEG-azide (MW 1000) is a biotin-labeled polyethylene glycol derivative bearing an azide functional group for bioconjugation. It is designed for applications such as PROTAC linker construction and protein labeling, enabling efficient attachment to alkyne- or strained-alkyne-containing molecules via click chemistry.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Provides biotin tag for affinity capture and detection.
  • PEG spacer increases solubility and reduces steric hindrance.
  • Typical purity 95% suitable for research applications.
Catalog No. 50-002-81632 Supplier Medchemexpress LLC Supplier No. HYW250928B100MG
Biotin-PEG-azide (MW 1000) is a biotin-labeled polyethylene glycol derivative bearing an azide functional group for bioconjugation. It is designed for applications such as PROTAC linker construction and protein labeling, enabling efficient attachment to alkyne- or strained-alkyne-containing molecules via click chemistry.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Provides biotin tag for affinity capture and detection.
  • PEG spacer increases solubility and reduces steric hindrance.
  • Typical purity 95% suitable for research applications.