Learn More
Medchemexpress LLC Biotin-PEG-azide | 956494-20-5 | MFCD21363314 | 98.7% | 2000.00 | Not specified - polydisperse PEG derivative (no fixed chemical formula) | 50 MG
SDP
Catalog No. 5000281245
Encompass_Preferred
Biotin-PEG-azide (MW 2000) is a biotin-labeled polyethylene glycol derivative with a terminal azide functional group for bioorthogonal conjugation. The reagent is supplied as a powder and is intended for use in click-chemistry bioconjugation workflows, enabling efficient biotinylation of alkyne-containing molecules and surfaces for downstream affinity capture and detection.
  • Contains terminal azide for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Peg spacer provides hydrophilicity and reduces nonspecific interactions.
  • Average molecular weight 2000 Da (polydisperse polymer).
  • High purity, suitable for research applications.
  • Stable as a powder at -20°C for long-term storage.
  • Useful for protein labeling, affinity capture, and general bioconjugation.
Catalog No. 50-002-81245 Supplier Medchemexpress LLC Supplier No. HYW250928C50MG
Biotin-PEG-azide (MW 2000) is a biotin-labeled polyethylene glycol derivative with a terminal azide functional group for bioorthogonal conjugation. The reagent is supplied as a powder and is intended for use in click-chemistry bioconjugation workflows, enabling efficient biotinylation of alkyne-containing molecules and surfaces for downstream affinity capture and detection.
  • Contains terminal azide for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Peg spacer provides hydrophilicity and reduces nonspecific interactions.
  • Average molecular weight 2000 Da (polydisperse polymer).
  • High purity, suitable for research applications.
  • Stable as a powder at -20°C for long-term storage.
  • Useful for protein labeling, affinity capture, and general bioconjugation.