Learn More
Medchemexpress LLC Biotin-PEG-biotin | 194920-55-3 | 99.3% | C26H44N6O6S2 | 10MG
SDP
Catalog No. 5000260341
Encompass_Preferred
Biotin-PEG-Biotin is a biotin-terminated polyethylene glycol (PEG) linker designed for use in PROTAC assembly and biotin-based affinity workflows. It provides dual biotin handles separated by a flexible PEG spacer, enabling conjugation, capture, and detection applications.
  • Biotinylated at both ends for affinity capture and pull-down workflows.
  • Peg spacer provides flexibility and improved solubility in polar organic solvents.
  • High purity for research applications - 99.3%.
  • Soluble in DMSO (~175 mg/mL); ultrasonic treatment may aid dissolution.
  • Store protected from light and under inert atmosphere; follow cold storage guidance.
Catalog No. 50-002-60341 Supplier Medchemexpress LLC Supplier No. HY140656A10MG
Biotin-PEG-Biotin is a biotin-terminated polyethylene glycol (PEG) linker designed for use in PROTAC assembly and biotin-based affinity workflows. It provides dual biotin handles separated by a flexible PEG spacer, enabling conjugation, capture, and detection applications.
  • Biotinylated at both ends for affinity capture and pull-down workflows.
  • Peg spacer provides flexibility and improved solubility in polar organic solvents.
  • High purity for research applications - 99.3%.
  • Soluble in DMSO (~175 mg/mL); ultrasonic treatment may aid dissolution.
  • Store protected from light and under inert atmosphere; follow cold storage guidance.