Learn More
Medchemexpress LLC Biotin-PEG-Biotin | 194920-55-3 | 99.3% | C26H44N6O6S2 | 25 MG
SDP
Catalog No. 5000352658
Encompass_Preferred
Biotin-PEG-Biotin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52658 Supplier Medchemexpress LLC Supplier No. 25MGHY140656A
Biotin-PEG-Biotin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.