Learn More
Medchemexpress LLC Biotin-PEG-Biotin | 194920-55-3 | 99.3% | C26H44N6O6S2 | 50 MG
SDP
Catalog No. 5000355467
Encompass_Preferred
Biotin-PEG-Biotin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-003-55467 Supplier Medchemexpress LLC Supplier No. HY140656A50MG
Biotin-PEG-Biotin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system to degrade target proteins