Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG-thiol (MW 2000) | 98.0% | 2000 (Average) | 100 MG
SDP

Catalog No. 5000360551
Encompass_Preferred

Biotin-PEG-Thiol (MW 2000) is an active compound. It is pegylated by binding to streptavidin or antibiotin with high affinity and specificity, allowing it to modify biomolecules, proteins, peptides, and other small molecule materials. This compound is widely used in the research of agent release and nano new materials.

  • Purity: 98.0%
  • Molecular weight: 2000 (Average)
  • Target: Fluorescent dye
  • Pathway: Others
  • In vitro solubility: DMSO: 125 mg/mL (Need ultrasonic)
  • Storage conditions: -20°C, protect from light, stored under nitrogen. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light, stored under nitrogen).

Catalog No. 50-003-60551 Supplier Medchemexpress LLC Supplier No. HY148263100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG-Thiol (MW 2000) is an active compound. It is pegylated by binding to streptavidin or antibiotin with high affinity and specificity, allowing it to modify biomolecules, proteins, peptides, and other small molecule materials. This compound is widely used in the research of agent release and nano new materials.

  • Purity: 98.0%
  • Molecular weight: 2000 (Average)
  • Target: Fluorescent dye
  • Pathway: Others
  • In vitro solubility: DMSO: 125 mg/mL (Need ultrasonic)
  • Storage conditions: -20°C, protect from light, stored under nitrogen. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light, stored under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.