Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG-Thiol (MW 2000) | 98.0% | 2000 (Average) | 25 MG
SDP

Catalog No. 5000362762
Encompass_Preferred

Biotin-PEG-Thiol (MW 2000) is an active compound that is pegylated by binding to streptavidin or antibiotin with high affinity and specificity. It can modify biomolecules, proteins, peptides, and other small molecule materials and is widely used in research concerning agent release and nano new materials.

  • Functions as an active compound.
  • Pegylated for high affinity and specificity binding to streptavidin or antibiotin.
  • Capable of modifying biomolecules, proteins, peptides, and other small molecule materials.
  • Widely used in research concerning agent release and nano new materials.
  • Solid, white to off-white appearance.
  • Store at -20°C, protected from light, and under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • Soluble in DMSO at 125 mg/mL (requires ultrasonication).

Catalog No. 50-003-62762 Supplier Medchemexpress LLC Supplier No. 25MGHY148263
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG-Thiol (MW 2000) is an active compound that is pegylated by binding to streptavidin or antibiotin with high affinity and specificity. It can modify biomolecules, proteins, peptides, and other small molecule materials and is widely used in research concerning agent release and nano new materials.

  • Functions as an active compound.
  • Pegylated for high affinity and specificity binding to streptavidin or antibiotin.
  • Capable of modifying biomolecules, proteins, peptides, and other small molecule materials.
  • Widely used in research concerning agent release and nano new materials.
  • Solid, white to off-white appearance.
  • Store at -20°C, protected from light, and under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • Soluble in DMSO at 125 mg/mL (requires ultrasonication).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.