Learn More
Medchemexpress LLC Biotin-PEG-Thiol (MW 2000) | 98.0% | 2000 (Average) | 50 MG
SDP
Catalog No. 5000363000
Encompass_Preferred
Biotin-PEG-Thiol (MW 2000) is an active compound that is pegylated by binding to streptavidin or antibiotin with high affinity and specificity. It can modify biomolecules, proteins, peptides, and other small molecule materials and is widely used in the research of agent release and nano new materials.
  • Active compound
  • Pegylated for high affinity and specificity binding to streptavidin or antibiotin
  • Modifies biomolecules, proteins, peptides, and other small molecule materials
  • Utilized in agent release and nano new materials research
  • Functions as a fluorescent dye
  • Acts as an inhibitor
Catalog No. 50-003-63000 Supplier Medchemexpress LLC Supplier No. HY14826350MG
Biotin-PEG-Thiol (MW 2000) is an active compound that is pegylated by binding to streptavidin or antibiotin with high affinity and specificity. It can modify biomolecules, proteins, peptides, and other small molecule materials and is widely used in the research of agent release and nano new materials.
  • Active compound
  • Pegylated for high affinity and specificity binding to streptavidin or antibiotin
  • Modifies biomolecules, proteins, peptides, and other small molecule materials
  • Utilized in agent release and nano new materials research
  • Functions as a fluorescent dye
  • Acts as an inhibitor