Learn More
Medchemexpress LLC Biotin-PEG1-azide | 1204085-48-2 | 99.9% | 356.44 g/mol | C14H24N6O3S | 10 MG
SDP
Catalog No. 5000204302
Encompass_Preferred
Biotin-PEG1-azide is a biotinylated polyethylene glycol (PEG1) azide reagent used as a cleavable linker and click chemistry handle for bioconjugation and antibody-drug conjugate synthesis. It provides an azide functional group for copper-catalyzed or strain-promoted azide-alkyne cycloaddition, enabling attachment to alkyne-containing partners.
  • Biotinylated PEG1 structure with terminal azide functional group.
  • Suitable for CuAAC and SPAAC click chemistry reactions.
  • High purity for research applications.
  • Oil appearance; store frozen for stability.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-04302 Supplier Medchemexpress LLC Supplier No. HYW09613310MG
Biotin-PEG1-azide is a biotinylated polyethylene glycol (PEG1) azide reagent used as a cleavable linker and click chemistry handle for bioconjugation and antibody-drug conjugate synthesis. It provides an azide functional group for copper-catalyzed or strain-promoted azide-alkyne cycloaddition, enabling attachment to alkyne-containing partners.
  • Biotinylated PEG1 structure with terminal azide functional group.
  • Suitable for CuAAC and SPAAC click chemistry reactions.
  • High purity for research applications.
  • Oil appearance; store frozen for stability.
  • Available in small milligram pack sizes for laboratory use.