Learn More
Medchemexpress LLC Biotin-PEG1-azide | 1204085-48-2 | 99.9% | 356.44 g/mol | C14H24N6O3S | 100 MG
SDP
Catalog No. 5000203083
Encompass_Preferred
Biotin-PEG1-azide is a biotin-labeled, single-unit polyethylene glycol (PEG1) azide reagent used for bioconjugation and as an ADC linker. The azide functional group enables click chemistry coupling, while the biotin tag allows affinity capture and detection.
  • Biotin-labeled PEG1 azide for affinity capture
  • Azide group for copper-catalyzed or strain-promoted click chemistry
  • High purity suitable for bioconjugation
  • Available as a 100 MG solid
  • Store solid at -20 °C; in solution store at -80 °C for long-term
Catalog No. 50-002-03083 Supplier Medchemexpress LLC Supplier No. HYW096133100MG
Biotin-PEG1-azide is a biotin-labeled, single-unit polyethylene glycol (PEG1) azide reagent used for bioconjugation and as an ADC linker. The azide functional group enables click chemistry coupling, while the biotin tag allows affinity capture and detection.
  • Biotin-labeled PEG1 azide for affinity capture
  • Azide group for copper-catalyzed or strain-promoted click chemistry
  • High purity suitable for bioconjugation
  • Available as a 100 MG solid
  • Store solid at -20 °C; in solution store at -80 °C for long-term