Learn More
Medchemexpress LLC Biotin-PEG1-azide | 1204085-48-2 | 99.9% | 356.44 g/mol | C14H24N6O3S | 50 MG
SDP
Catalog No. 5000204301
Encompass_Preferred
Biotin-PEG1-azide is a biotinylated polyethylene glycol (PEG1) azide reagent used as a linker and click-chemistry handle for conjugation, labeling, and affinity-based capture in research applications.
  • introduces a biotin tag and an azide functional group for covalent conjugation
  • suitable for copper-catalyzed and strain-promoted azide-alkyne cycloaddition
  • high purity for reliable experimental results
  • available in small-scale quantities suitable for synthesis and labeling workflows
  • storage recommended at low temperatures to preserve stability
Catalog No. 50-002-04301 Supplier Medchemexpress LLC Supplier No. HYW09613350MG
Biotin-PEG1-azide is a biotinylated polyethylene glycol (PEG1) azide reagent used as a linker and click-chemistry handle for conjugation, labeling, and affinity-based capture in research applications.
  • introduces a biotin tag and an azide functional group for covalent conjugation
  • suitable for copper-catalyzed and strain-promoted azide-alkyne cycloaddition
  • high purity for reliable experimental results
  • available in small-scale quantities suitable for synthesis and labeling workflows
  • storage recommended at low temperatures to preserve stability