Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG1-NH2 | 811442-85-0 | 97.0% | 330.45 g/mol | C14H26N4O3S | 100 MG
SDP

Catalog No. 5000202645
Encompass_Preferred

Biotin-PEG1-NH2 is a cleavable one-unit polyethylene glycol (PEG) linker bearing a biotin tag and a terminal primary amine (-NH2), designed for use in the synthesis of antibody-drug conjugates (ADCs). Intended for research use only.

  • Cleavable 1-unit PEG linker suitable for ADC synthesis.
  • Biotin tag for affinity capture or detection.
  • Terminal primary amine (-NH2) for conjugation chemistries.
  • Purity 97.0%.
  • Molecular weight 330.45 g/mol.
  • CAS number 811442-85-0.
  • Form: white to off-white solid; store protected from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Available packaging: 25 MG, 50 MG, 100 MG, 200 MG (quote), 500 MG (quote).

Catalog No. 50-002-02645 Supplier Medchemexpress LLC Supplier No. HYW096135100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG1-NH2 is a cleavable one-unit polyethylene glycol (PEG) linker bearing a biotin tag and a terminal primary amine (-NH2), designed for use in the synthesis of antibody-drug conjugates (ADCs). Intended for research use only.

  • Cleavable 1-unit PEG linker suitable for ADC synthesis.
  • Biotin tag for affinity capture or detection.
  • Terminal primary amine (-NH2) for conjugation chemistries.
  • Purity 97.0%.
  • Molecular weight 330.45 g/mol.
  • CAS number 811442-85-0.
  • Form: white to off-white solid; store protected from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Available packaging: 25 MG, 50 MG, 100 MG, 200 MG (quote), 500 MG (quote).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.