Learn More
Medchemexpress LLC Biotin-PEG10-NHS ester | 98.0% | C37H64N4O16S | 25 MG
SDP
Catalog No. 5000354704
Encompass_Preferred
This PEG-based linker is utilized in the synthesis of degraders, specifically within the PROTAC (Proteolysis Targeting Chimeras) modality. PROTACs are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. They function by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • Intended for research use only
Catalog No. 50-003-54704 Supplier Medchemexpress LLC Supplier No. 25MGHY143855
This PEG-based linker is utilized in the synthesis of degraders, specifically within the PROTAC (Proteolysis Targeting Chimeras) modality. PROTACs are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. They function by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • Intended for research use only