Learn More
Medchemexpress LLC Biotin-PEG11-amine | 1418022-42-0 | 99.9% | C34H66N4O13S | 100 MG
SDP
Catalog No. 5000355002
Encompass_Preferred
Biotin-PEG11-amine is a biotin-labeled, PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs selectively degrade target proteins
  • PROTACs utilize the ubiquitin-proteasome system
Catalog No. 50-003-55002 Supplier Medchemexpress LLC Supplier No. HY140900100MG
Biotin-PEG11-amine is a biotin-labeled, PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs selectively degrade target proteins
  • PROTACs utilize the ubiquitin-proteasome system