Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG11-amine | 1418022-42-0 | MFCD21363309 | 99.9% | C34H66N4O13S | 10MG
SDP

Catalog No. 5000260866
Encompass_Preferred

Biotin-PEG11-amine is a biotin-labeled polyethylene glycol linker with a terminal primary amine, supplied as a solid for research use. It is intended for biotinylation and incorporation into PROTACs or other conjugates, offering improved aqueous solubility and a flexible spacer to reduce steric hindrance during coupling reactions.

  • Provides a terminal primary amine for coupling to carboxyl or phosphate groups.
  • Contains an 11-unit PEG spacer to improve solubility and flexibility.
  • Supports biotin-based affinity capture and detection workflows.
  • Supplied as a white to off-white solid with high purity.
  • Recommended storage: -20°C protected from light; in solvent, -80°C for long-term storage.

Catalog No. 50-002-60866 Supplier Medchemexpress LLC Supplier No. HY14090010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG11-amine is a biotin-labeled polyethylene glycol linker with a terminal primary amine, supplied as a solid for research use. It is intended for biotinylation and incorporation into PROTACs or other conjugates, offering improved aqueous solubility and a flexible spacer to reduce steric hindrance during coupling reactions.

  • Provides a terminal primary amine for coupling to carboxyl or phosphate groups.
  • Contains an 11-unit PEG spacer to improve solubility and flexibility.
  • Supports biotin-based affinity capture and detection workflows.
  • Supplied as a white to off-white solid with high purity.
  • Recommended storage: -20°C protected from light; in solvent, -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.