Learn More
Medchemexpress LLC Biotin-PEG11-Mal | 1646619-56-8 | 96.5% | C41H71N5O16S | 25 MG
SDP
Catalog No. 5000357373
Encompass_Preferred
Biotin-PEG11-Mal is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Functions by harnessing the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57373 Supplier Medchemexpress LLC Supplier No. 25MGHY140909
Biotin-PEG11-Mal is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Functions by harnessing the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins