Learn More
Medchemexpress LLC Biotin-PEG11-mal | 1646619-56-8 | 98.1% | 922.09 g·mol⁻¹ | C41H71N5O16S | 5 MG
SDP
Catalog No. 5000222164
Encompass_Preferred
Biotin-PEG11-Mal is a biotinylated, maleimide-terminated polyethylene glycol (PEG11) linker supplied as a research reagent for conjugation and PROTAC synthesis. The maleimide moiety allows selective coupling to thiol groups, while the biotin tag enables affinity capture or detection. The product is provided as a purified solid with high reported purity suitable for synthetic and biochemical workflows.
  • Maleimide functional group enables thiol-selective conjugation to cysteine residues.
  • Biotin tag enables affinity capture and detection in pull-down and labeling assays.
  • PEG11 spacer improves solubility and provides a flexible linker between moieties.
  • High reported purity (98.12%) suitable for synthetic applications.
  • Offered in small laboratory pack sizes for convenient research use.
Catalog No. 50-002-22164 Supplier Medchemexpress LLC Supplier No. HY1409095MG
Biotin-PEG11-Mal is a biotinylated, maleimide-terminated polyethylene glycol (PEG11) linker supplied as a research reagent for conjugation and PROTAC synthesis. The maleimide moiety allows selective coupling to thiol groups, while the biotin tag enables affinity capture or detection. The product is provided as a purified solid with high reported purity suitable for synthetic and biochemical workflows.
  • Maleimide functional group enables thiol-selective conjugation to cysteine residues.
  • Biotin tag enables affinity capture and detection in pull-down and labeling assays.
  • PEG11 spacer improves solubility and provides a flexible linker between moieties.
  • High reported purity (98.12%) suitable for synthetic applications.
  • Offered in small laboratory pack sizes for convenient research use.