Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG11-SH | 1650579-23-9 | 98.0% | C34H65N3O13S2 | 100 MG
SDP

Catalog No. 5000355291
Encompass_Preferred

Biotin-PEG11-SH enables biotin labeling of protein. It is intended for research use only and is not sold to patients. This product appears as an off-white to light yellow solid with a molecular weight of 788.02. It can be stored at -20°C under nitrogen or at -80°C for 6 months when in solvent.

  • Enables biotin labeling of protein
  • For research use only
  • Appears as an off-white to light yellow solid
  • Molecular weight: 788.02
  • Can be stored at -20°C under nitrogen
  • Soluble in DMSO at concentrations of ≥ 100 mg/mL

Catalog No. 50-003-55291 Supplier Medchemexpress LLC Supplier No. HY145445100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG11-SH enables biotin labeling of protein. It is intended for research use only and is not sold to patients. This product appears as an off-white to light yellow solid with a molecular weight of 788.02. It can be stored at -20°C under nitrogen or at -80°C for 6 months when in solvent.

  • Enables biotin labeling of protein
  • For research use only
  • Appears as an off-white to light yellow solid
  • Molecular weight: 788.02
  • Can be stored at -20°C under nitrogen
  • Soluble in DMSO at concentrations of ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.