Learn More
Medchemexpress LLC Biotin-PEG11-SH | 1650579-23-9 | 98.0% | C34H65N3O13S2 | 50 MG
SDP
Catalog No. 5000355776
Encompass_Preferred
BIOTIN-PEG11-SH is a chemical reagent that enables the biotin labeling of proteins. This compound is provided as an off-white to light yellow solid with a purity of 98.0% and is suitable for various research applications.
  • Enables biotin labeling of proteins
  • For research use only
  • Off-white to light yellow solid appearance
  • Classified as a fluorescent dye
  • Used in labeling chemistry
Catalog No. 50-003-55776 Supplier Medchemexpress LLC Supplier No. HY14544550MG
BIOTIN-PEG11-SH is a chemical reagent that enables the biotin labeling of proteins. This compound is provided as an off-white to light yellow solid with a purity of 98.0% and is suitable for various research applications.
  • Enables biotin labeling of proteins
  • For research use only
  • Off-white to light yellow solid appearance
  • Classified as a fluorescent dye
  • Used in labeling chemistry