Learn More
Medchemexpress LLC Biotin-PEG12-NHS ester | 1934296-88-4 | 99.3% | C41H72N4O18S | 100 MG
SDP
Catalog No. 5000355406
Encompass_Preferred
Biotin-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. This compound utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-003-55406 Supplier Medchemexpress LLC Supplier No. HY140892100MG
Biotin-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. This compound utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • For research use only