Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG12-NHS ester | 1934296-88-4 | 99.3% | C41H72N4O18S | 50 MG
SDP

Catalog No. 5000356638
Encompass_Preferred

Biotin-PEG12-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a white to off-white oil with a purity of 99.31%.

  • Purity of 99.31%
  • Molecular weight of 941.09
  • Chemical formula: C41H72N4O18S
  • Appears as a white to off-white oil
  • Recommended storage for pure form at -20°C for 3 years
  • Recommended storage in solvent at -80°C for 6 months, or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
  • Useful as a PROTAC linker for degrading target proteins

Catalog No. 50-003-56638 Supplier Medchemexpress LLC Supplier No. HY14089250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG12-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a white to off-white oil with a purity of 99.31%.

  • Purity of 99.31%
  • Molecular weight of 941.09
  • Chemical formula: C41H72N4O18S
  • Appears as a white to off-white oil
  • Recommended storage for pure form at -20°C for 3 years
  • Recommended storage in solvent at -80°C for 6 months, or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
  • Useful as a PROTAC linker for degrading target proteins

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.