Learn More
Medchemexpress LLC Biotin-PEG12-NHS ester | 1934296-88-4 | 99.31% | C41H72N4O18S | 25 MG
SDP
Catalog No. 5000352666
Encompass_Preferred
Biotin-PEG12-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52666 Supplier Medchemexpress LLC Supplier No. 25MGHY140892
Biotin-PEG12-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins