Learn More
Broadpharm Biotin-PEG12-NHS ester | 365441-71-0 | MFCD13184968 | 250 mg
Catalog No. NC1096571
Encompass
Biotin-PEG12-NHS ester | Purity: 98% | Mol Wt: 941.1 | 365441-71-0 | MFCD13184968 | 250 mg
Catalog No. NC1096571 Supplier Broadpharm Supplier No. BP21619250MG
Add to Cart
Add to Cart
Biotin-PEG12-NHS ester | Purity: 98% | Mol Wt: 941.1 | 365441-71-0 | MFCD13184968 | 250 mg