Learn More
TARGETMOL CHEMICALS INC Biotin-PEG12-NHS ester 50MG
SDP
Catalog No. 502878396
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Biotin-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. Purity ≥98%
Catalog No. 50-287-8396 Supplier TARGETMOL CHEMICALS INC Supplier No. T1756150MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Biotin-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. Purity ≥98%