Learn More
Medchemexpress LLC Biotin-PEG2-acid | 1365655-89-5 | MFCD22056305 | ≥97.0% | 403.49 g·mol⁻¹ | C17H29N3O6S | 250 MG
SDP
Catalog No. 5000204098
Encompass_Preferred
Biotin-PEG2-acid is a biotinylated polyethylene glycol linker with a terminal carboxylic acid, used as a non-cleavable ADC and PROTAC linker for bioconjugation, affinity labeling, and linker synthesis. It is supplied in small laboratory pack sizes for conjugation workflows.
  • Biotinylated PEG linker with a terminal carboxylic acid for conjugation.
  • Non-cleavable two-unit PEG spacer suited for ADC and PROTAC syntheses.
  • High purity (≥97.0%) for reliable conjugation performance.
  • Molecular weight 403.49 g·mol⁻¹; formula C17H29N3O6S.
  • Available in small-scale quantities such as 250 MG.
  • Store protected from light under argon at -20°C; in solvent store at -80°C for long-term.
Catalog No. 50-002-04098 Supplier Medchemexpress LLC Supplier No. HY126958250MG
Biotin-PEG2-acid is a biotinylated polyethylene glycol linker with a terminal carboxylic acid, used as a non-cleavable ADC and PROTAC linker for bioconjugation, affinity labeling, and linker synthesis. It is supplied in small laboratory pack sizes for conjugation workflows.
  • Biotinylated PEG linker with a terminal carboxylic acid for conjugation.
  • Non-cleavable two-unit PEG spacer suited for ADC and PROTAC syntheses.
  • High purity (≥97.0%) for reliable conjugation performance.
  • Molecular weight 403.49 g·mol⁻¹; formula C17H29N3O6S.
  • Available in small-scale quantities such as 250 MG.
  • Store protected from light under argon at -20°C; in solvent store at -80°C for long-term.