Learn More
Medchemexpress LLC Biotin-PEG2-aldehyde | 00-00-0 | 95.0% | 387.5 g/mol | C17H29N3O5S | 5 MG
SDP
Catalog No. 5000262695
Encompass_Preferred
Biotin-PEG2-aldehyde is a biotinylated PEG linker bearing a terminal aldehyde functional group for bioconjugation and PROTAC synthesis. The short PEG2 spacer increases aqueous solubility and provides a flexible linkage between the biotin tag and attached payloads.
  • Terminal aldehyde enables chemoselective conjugation (hydrazone, oxime, reductive amination)
  • PEG2 spacer improves aqueous solubility and flexibility
  • Biotin tag enables affinity capture with streptavidin-based reagents
  • Compatible with PROTAC and probe synthesis workflows
  • High purity suitable for research use (95.0%)
  • Available in small research-friendly pack sizes
Catalog No. 50-002-62695 Supplier Medchemexpress LLC Supplier No. HY1334565MG
Biotin-PEG2-aldehyde is a biotinylated PEG linker bearing a terminal aldehyde functional group for bioconjugation and PROTAC synthesis. The short PEG2 spacer increases aqueous solubility and provides a flexible linkage between the biotin tag and attached payloads.
  • Terminal aldehyde enables chemoselective conjugation (hydrazone, oxime, reductive amination)
  • PEG2 spacer improves aqueous solubility and flexibility
  • Biotin tag enables affinity capture with streptavidin-based reagents
  • Compatible with PROTAC and probe synthesis workflows
  • High purity suitable for research use (95.0%)
  • Available in small research-friendly pack sizes