Learn More
Medchemexpress LLC Biotin-PEG2-aldehyde | 00-00-0 | 95.0% | C17H29N3O5S | 10MG
SDP
Catalog No. 5000261864
Encompass_Preferred
Biotin-PEG2-aldehyde is a PEG-based linker bearing a biotin affinity tag and an aldehyde functional group, supplied as a solid reagent for PROTAC synthesis and general bioconjugation. The aldehyde enables chemoselective ligations while the PEG2 spacer improves solubility and reduces steric hindrance.
Catalog No. 50-002-61864 Supplier Medchemexpress LLC Supplier No. HY13345610MG
Biotin-PEG2-aldehyde is a PEG-based linker bearing a biotin affinity tag and an aldehyde functional group, supplied as a solid reagent for PROTAC synthesis and general bioconjugation. The aldehyde enables chemoselective ligations while the PEG2 spacer improves solubility and reduces steric hindrance.