Learn More
Medchemexpress LLC Biotin-PEG2-alkyne | 2227450-68-0 | MFCD31922733 | 95.6% | 369.48 g/mol | C17H27N3O4S | 1 G
SDP
Catalog No. 5000284248
Encompass_Preferred
Biotin-PEG2-alkyne is a short polyethylene glycol (PEG2) linker that bears a biotin group and a terminal alkyne. It is used to biotinylate azide-containing molecules via copper(I)-catalyzed azide-alkyne cycloaddition and for affinity enrichment workflows.
  • Enables copper(I)-catalyzed azide-alkyne cycloaddition for click chemistry labeling.
  • Provides a biotin tag compatible with streptavidin-based affinity capture and enrichment.
  • Short PEG2 spacer minimizes steric hindrance while improving solubility.
  • Off-white to light yellow solid, typically supplied with high purity for research use.
  • Available in multiple pack sizes to support small-scale labeling and larger preparations.
Catalog No. 50-002-84248 Supplier Medchemexpress LLC Supplier No. HYW6150951G
Biotin-PEG2-alkyne is a short polyethylene glycol (PEG2) linker that bears a biotin group and a terminal alkyne. It is used to biotinylate azide-containing molecules via copper(I)-catalyzed azide-alkyne cycloaddition and for affinity enrichment workflows.
  • Enables copper(I)-catalyzed azide-alkyne cycloaddition for click chemistry labeling.
  • Provides a biotin tag compatible with streptavidin-based affinity capture and enrichment.
  • Short PEG2 spacer minimizes steric hindrance while improving solubility.
  • Off-white to light yellow solid, typically supplied with high purity for research use.
  • Available in multiple pack sizes to support small-scale labeling and larger preparations.